Documenti fondamentali
Z243280
ACE Squibb separatory funnel with Locktight stopper
capacity 500 mL, stopper joint: ST/NS 24/40
Scegli un formato
1 EA
CHF 378.00
CHF 378.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 378.00
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
CHF 378.00
Check Cart for Availability
Giunto
joint: ST/NS stopper24/40
Produttore/marchio commerciale
Ace Glass 723161
Capacità
500 mL
Categorie correlate
Descrizione generale
European style funnel with new leaktight locking stopper that will not fall out during shaking or transportation. The stopper is secured into a screw-threaded top joint via cap and Viton® O-ring for a positive, leaktight seal. Supplied complete with 1:5 PTFE plug, stopper, cap, and O-ring.
Note legali
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)