Documenti fondamentali
Z177865
Scienceware® Ellipso-Spoon® sampler
L 21 cm
Sinonimo/i:
curved laboratory spatula, laboratory spoon
Scegli un formato
1 EA
CHF 43.90
CHF 43.90
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 43.90
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
CHF 43.90
Check Cart for Availability
Produttore/marchio commerciale
Bel-Art F368070021
Lunghezza
21 cm
Descrizione generale
Special curved shape permits contact with all corners of vessels, bottles, or containers. No corner out of reach and no loss of materials. Handles crystals, powders, granulated and solid substances, even thick liquids or delicate substances. SS, will not bend. Has both spoon end and spatula end.
Note legali
Ellipso-Spoon is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)