Z167525
Aldrich® Schlenk-type Liebig condenser
male joint: ST/NS 14/20, female joint: ST/NS 14/20
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Etichetta
P
Giunto
female joint: ST/NS14/20
male joint: ST/NS14/20
Lunghezza
100 mm
Descrizione generale
Replacement part for Aldrich Air-Sensitive glass kit. Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Articoli
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.