Documenti fondamentali
Z126772
Sigma-Aldrich
Standard point hypodermic needles, 304 stainless steel
gauge 12, L 2 in.
Scegli un formato
12 EA
CHF 499.00
CHF 499.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
12 EA
CHF 499.00
About This Item
Codice UNSPSC:
42142523
NACRES:
NB.03
CHF 499.00
Check Cart for Availability
Materiali
304 stainless steel needle
chrome-plated hub
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Popper 7232
Lunghezza
2 in.
Gauge
12
Descrizione generale
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)