Documenti
TEM-FF400CU
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 400 mesh, box of 25
Sinonimo/i:
FORMVAR FILM 400# COPPER 25/PK
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
box of 25
Misura
400 mesh
Descrizione generale
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 400 mesh. All of our film is laid on the shiny side of the grid.