Documenti fondamentali
TEM-71150
Sigma-Aldrich
Grid Storage Box
capacity, 50 (wells), pkg of 1 ea
Sinonimo/i:
GRID STORAGE BOX 50-CAP CLEAR COVER R13A2
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Caratteristiche
capacity 50 (wells)
Confezionamento
pkg of 1 ea
Descrizione generale
Grid Storage Box, 50 CapacityStorage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)