Documenti fondamentali
SYNV165045
Synthware vacuum trap with serrated hose connections
joint: ST/NS 45/50, body O.D. 50 mm, L 250 mm , below joint
Sinonimo/i:
Synthware V165045
Scegli un formato
1 EA
CHF 173.00
CHF 173.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 173.00
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
CHF 173.00
Check Cart for Availability
Giunto
joint: ST/NS45/50
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware V165045
Lunghezza
250 mm , below joint
Diam. est. corpo
50 mm
Descrizione generale
With joints for easy removal of condensate. Serrated hose connections for side tube have an O.D. of 13 mm at the smallest serration and 16 mm at the largest serration. Center tube hose connection is always 10mm for easy connection to manifold bank tubes which are also with 10 mm hose connection.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 1
1 of 1
Questions
  1. Hello, I just want to make sure if the trap comes with every part : the head and the body. Thanks in advance
    1 answer
    1. The head and body are included with this product, as indicated in the product image.