Documenti fondamentali
SYNC204100
Synthware Liebig condenser with 8 mm OD hose connections
joint: ST/NS 24/40, jacket L 100 mm
Sinonimo/i:
Synthware C204100, Straight condenser
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C204100
Lunghezza camicia
100 mm
Descrizione generale
Synthware Liebig condenser features a 100 mm jacketed length for superior cooling efficiency. With standard taper outer joint at the top (24/40) and a lower inner drip tip joint. Made from durable borosilicate glass.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)