Documenti
SLW1060/14D
Pyrex® Watch glasses
nominal diameter 100 mm
Sinonimo/i:
Pyrex, SciLabware 1060/14D
About This Item
Codice UNSPSC:
41121801
NACRES:
NB.43
Confezionamento
pkg of 10
Produttore/marchio commerciale
1060/04D
SciLabware
Diametro nominale
100 mm
Descrizione generale
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Note legali
PYREX is a registered trademark of Corning, Inc.