Documenti fondamentali
SLW1000/28D
Pyrex® Griffin beakers, low form, with printed trace code
2000 mL
Sinonimo/i:
Pyrex, SciLabware 1000/28D
Scegli un formato
10 EA
CHF 324.00
CHF 324.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 324.00
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
CHF 324.00
Check Cart for Availability
Materiali
borosilicate glass
white graduation
Caratteristiche
low form
with printed trace code
Produttore/marchio commerciale
1000/28D
SciLabware
Alt. × Diam.
185 mm × 130 mm
Capacità
2000 mL
Descrizione generale
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White graduations and marking spot
Altre note
Browse all glass beakers
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)