Documenti fondamentali
R456217
N-(3-CYANO-BENZO(4,5)IMIDAZO(1,2-A)PYRIMIDIN-2-YL)-3-MEO-4-PENTYLOXY-BENZAMIDE
AldrichCPR
Scegli un formato
25MG
CHF 34.40
CHF 34.40
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
25MG
CHF 34.40
About This Item
Formula condensata:
C24H23N5O3
Numero CAS:
Peso molecolare:
429.48
Numero MDL:
CHF 34.40
Check Cart for Availability
Altre note
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)