GF55393847
Nickel/Cobalt
powder, Ni 97%/Co 3%, mean particle size 50 micron, weight 20 g, condition gas atomized
Sinonimo/i:
NE296011
About This Item
Prodotti consigliati
Forma fisica
powder
Produttore/marchio commerciale
Goodfellow 553-938-47
Peso
20 g
Dimens. media particelle
50 μm