DWK218666901
DURAN® wide mouth amber laboratory bottle
without cap, round amber glass bottle, bottle capacity (3,500 mL), non-sterile
Sinonimo/i:
218666901
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Materiali
round amber glass bottle
Sterilità
non-sterile
Caratteristiche
without cap
Confezionamento
case of 1 ea
Produttore/marchio commerciale
DURAN® 218666901
Capacità bottiglia
(3,500 mL)
Raccordi
GL 80 cap
GL 80 neck
Descrizione generale
UV protection up to approx. 500 nm due to external colouration. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties. Special thread design for rapid opening and closing in less than a turn Optimum filling and pouring and particularly easy cleaning. Spoons, spatulas and tweezers can be manipulated with ease.
Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base areaEasy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic print.
Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base areaEasy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic print.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)