CLS762422
Pyrex® HDPE stopper, improved form
stopper joint: 22
Sinonimo/i:
flask stopper, flask stoppers, stoppers, Corning® 7624
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.43
Prodotti consigliati
Giunto
joint: stopper22
Produttore/marchio commerciale
Corning 7624-22
Categorie correlate
Descrizione generale
Blue and white stoppers with closed bottoms, made of linear high-density polyethylene. Improved design fully protects the neck in the event of accidental tipping.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)