Documenti fondamentali
CLS316610
Pyrex® cloning cylinder
O.D. × H 10 mm × 10 mm
Sinonimo/i:
lab equipment accessory
Scegli un formato
125 EA
CHF 94.20
CHF 94.20
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
125 EA
CHF 94.20
About This Item
Codice UNSPSC:
41106618
NACRES:
NB.43
CHF 94.20
Check Cart for Availability
Materiali
borosilicate glass
cylindrical
Sterilità
non-sterile
Confezionamento
pack of 125
Produttore/marchio commerciale
Corning 3166-10
D.E. × alt.
10 mm × 10 mm
Descrizione generale
Clone a single cell or group of cells by surrounding them with this Pyrex glass open ended cloning cylinder. Dip the end of the cylinder into sterile silicone grease before pressing to the bottom of a culture dish to create an isolated well. Cylinders are autoclavable or can be dry heat sterilized.
Caratteristiche e vantaggi
  • Pyrex® cloning cylinder Type I Class A - Low-Expansion Borosilicate Glass
  • Compatible with heating using hot plate, flame, oven, microwave, or heating mantles.
  • Product can be cooled to -230°C.
  • Product Dimensions - 10 mm OD x 10 mm High.
  • DO NOT plunge product directly into a liquid coolant.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)