Documenti fondamentali
CAD6155
1-way tubing connector
ML to hose end for 1/16 in. to 3/32 in. I.D. tubing (plated brass)
Scegli un formato
10 EA
CHF 104.00
CHF 104.00
Spedizione prevista il05 giugno 2025
Scegli un formato
Cambia visualizzazione
10 EA
CHF 104.00
About This Item
Codice UNSPSC:
40183110
NACRES:
NB.03
CHF 104.00
Spedizione prevista il05 giugno 2025
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
Cadence Science 6155
Categorie correlate
Descrizione generale
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Note legali
Micro-Mate is a registered trademark of Cadence Science
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)