Documenti
BR90138
BRAND® Griffin beaker with spout
low form, PFA, volume 100 mL
Sinonimo/i:
Beakers, PFA, Plastics
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Materiali
PFA
Caratteristiche
graduations
low form
Produttore/marchio commerciale
BRAND 90138
Volume
100 mL
Descrizione generale
The BRAND® Griffin Beaker with Spout is crafted from high-quality ETFE, offering excellent chemical resistance and durability for laboratory applications. This translucent beaker features clear graduation markings for precise measurement, allowing for accurate liquid handling. The beaded rim enhances strength and provides a comfortable grip, while the integrated spout facilitates easy pouring, minimizing spills and improving accuracy during liquid transfer. Ideal for a variety of experiments, this beaker combines functionality with reliability, making it an essential tool for any laboratory setting.
Altre note
Browse all plastic beakers
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG