Documenti fondamentali
BR780740
BRAND® screw cap for micro tubes
white
Sinonimo/i:
Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps, Tubes
Scegli un formato
1000 EA
CHF 127.00
CHF 127.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1000 EA
CHF 127.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
CHF 127.00
Check Cart for Availability
Materiali
polypropylene
white
Confezionamento
pack of 1000 ea
Produttore/marchio commerciale
BRAND 780740
Descrizione generale
The BRAND® Screw Cap for Micro Tubes is designed specifically for use with micro tubes 780730 - 780734 and 780750 - 780754. Made from high-quality polypropylene (PP), these screw caps feature a silicone seal that ensures a secure closure, preventing leaks and contamination. They are suitable for a wide range of applications, withstanding temperatures from -196 °C to +121 °C. Certified as BIO-CERT® PCR QUALITY, these caps are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, making them ideal for sensitive molecular biology applications.
Note legali
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)