Documenti fondamentali
BR74506
BRAND® filter funnel support with base plate
for 2 funnels
Sinonimo/i:
Accessories, Filter funnels, Funnels, Supports
Scegli un formato
1 EA
CHF 182.00
CHF 182.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 182.00
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.24
CHF 182.00
Check Cart for Availability
Materiali
polypropylene
Caratteristiche
for 2 funnels
Produttore/marchio commerciale
BRAND 74506
Lungh. × largh.
250 mm × 140 mm
Descrizione generale
These PP plates feature a stainless steel support rod with dimensions of 12 mm in diameter and 595 mm in length. They are designed to support 2 or 4 funnels with a top outer diameter ranging from 50 mm to 120 mm. The height of the funnels is adjustable, allowing for customized setups based on your specific needs.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)