Documenti fondamentali
BR718620
BRAND® counting chamber BLAUBRAND® Neubauer pattern
with clips, double ruled
Sinonimo/i:
BLAUBRAND®, Cell culture, Counting Chambers, Neubauer
Scegli un formato
1 EA
CHF 95.60
CHF 95.60
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 95.60
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
CHF 95.60
Check Cart for Availability
Materiali
colorless
glass
Sterilità
non-sterile
Caratteristiche
with clips
Produttore/marchio commerciale
BRAND 718620
Dimens. camera
0.1 mm , depth
Pozzetti
1
Descrizione generale
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling shows 9 large squares of 1 mm² each.The four large squares in the corners marked "L" for counting leucocytes are each subdivided into 16 squares with 0.25 mm sides.The large square in the center is subdivided into 16 squares with 0.25 mm sides. Each group square consists of 16 mini squares with 0.05 mm sides, each having an area of 0.0025 mm². The 5 group squares in the large central square are used for counting thrombocytes and erythrocytes.Contrary to the more advanced Improved Neubauer counting chambers, the counting area of each group square is limited by the outer one of the triple boundary lines.
Caratteristiche e vantaggi
  • BLAUBRAND® quality
  • Outstanding measurement precision
  • 100% certified quality
Confezionamento
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)