Documenti fondamentali
BR43305
BRAND® BLAUBRAND® density bottle, Gay-Lussac pattern, calibrated
capacity 5 mL, borosilicate glass
Sinonimo/i:
BLAUBRAND®, Gay-Lussac
Scegli un formato
1 EA
CHF 49.40
CHF 49.40
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 49.40
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.71
CHF 49.40
Check Cart for Availability
Materiali
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Giunto
joint ST/NS neck10/19
Caratteristiche
with cap
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 43305
Capacità
5 mL
Descrizione generale
DIN ISO 3507, type Gay-Lussac. Calibrated to contain (TC, In). Incl. individual certificate. Stopper NS 10/19 with capillary. Top of stopper ground and polished. The volume in cm³ is specified to a precision of 3 decimal places.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.