BR36984
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 2000 mL, neck joint: ST/NS 29/32, glass stopper
Sinonimo/i:
BLAUBRAND®, USP, Volumetric flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
glass stopper
Giunto
joint: ST/NS neck29/32
Caratteristiche
accuracy class A
Confezionamento
pack of 2
Produttore/marchio commerciale
BRAND 36984
Capacità
2000 mL
Categorie correlate
Descrizione generale
The BRAND® volumetric flask BLAUBRAND® USP conformity certified is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for a variety of laboratory applications. Each flask comes with a USP batch certificate, confirming its adherence to stringent quality and safety standards. This volumetric flask is an essential tool for precise analytical work, making it ideal for laboratories that require reliable and certified liquid handling solutions.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)