Principaux documents
V900282
Sigma-Aldrich
Cyclohexanone
Vetec, reagent grade, 98%
About This Item
Formule linéaire :
C6H10(=O)
Numéro CAS:
Poids moléculaire :
98.14
Beilstein:
385735
Numéro CE :
Numéro MDL:
Code UNSPSC :
12352100
ID de substance PubChem :
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Qualité
reagent grade
Densité de vapeur
3.4 (vs air)
Pression de vapeur
3.4 mmHg ( 20 °C)
Gamme de produits
Vetec
Essai
98%
Forme
liquid
Température d'inflammation spontanée
788 °F
Limite d'explosivité
1.1 %, 100 °F
9.4 %
Indice de réfraction
n20/D 1.450 (lit.)
pH
~7 (20 °C, 70 g/L)
pb
155 °C (lit.)
Pf
−47 °C (lit.)
Densité
0.947 g/mL at 25 °C (lit.)
Chaîne SMILES 
O=C1CCCCC1
InChI
1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2
Clé InChI
JHIVVAPYMSGYDF-UHFFFAOYSA-N
Informations légales
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Pictogrammes
FlameCorrosionExclamation mark
Mention d'avertissement
Danger
Classification des risques
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Organes cibles
Respiratory system
Code de la classe de stockage
3 - Flammable liquids
Classe de danger pour l'eau (WGK)
WGK 1
Point d'éclair (°F)
111.2 °F - closed cup
Point d'éclair (°C)
44 °C - closed cup
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2629-2637 (2012-09-07)
Iron (and to a lesser extent manganese) in the wall of a 316 stainless steel (SS) reactor is responsible for the hydrogenation of cyclohexanone to cyclohexanol when using an aqueous formic acid solution under high temperature and pressure water (HTPW)
Application of fragment screening and merging to the discovery of inhibitors of the Mycobacterium tuberculosis cytochrome P450 CYP121.
Sean A Hudson et al.
Angewandte Chemie (International ed. in English), 51(37), 9311-9316 (2012-08-15)
Brahm J Yachnin et al.
Journal of the American Chemical Society, 134(18), 7788-7795 (2012-04-18)
The Baeyer-Villiger monooxygenases (BVMOs) are a family of bacterial flavoproteins that catalyze the synthetically useful Baeyer-Villiger oxidation reaction. This involves the conversion of ketones into esters or cyclic ketones into lactones by introducing an oxygen atom adjacent to the carbonyl
Alakananda Hajra et al.
Organic letters, 14(21), 5488-5491 (2012-10-18)
Dehydrogenative aromatization of cyclohexanone imines to arylamines has been achieved using a palladium catalyst under aerobic conditions. The reaction is applicable to a variety of imines that are either preformed or generated in situ from cyclohexanone derivatives and aryl or
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2638-2643 (2012-09-07)
The processing of renewable feedstocks to platform chemicals and, to a lesser degree, fuels is a key part of sustainable development. In particular, the combination of lignocellulosic biomass with hydrothermal upgrading (HTU), using high temperature and pressure water (HTPW), is