Principaux documents
Z740155
LifeSep magnetic separation stand
1.5S, for 1.5 mL tubes
Synonyme(s) :
magnetic separation equipment
4 EA
264.00 CHF
264.00 CHF
Date d'expédition estimée le31 mars 2025
Changer de vue
4 EA
264.00 CHF
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.56
264.00 CHF
Date d'expédition estimée le31 mars 2025
Caractéristiques
for 1.5 mL tubes
Fabricant/nom de marque
Dexter Magnetic Technologies 2501009
Description générale
Magnetic separators for single tubes are durable and easy-to-use. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Open-faced design for clear visibility
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
  • Durable and easy to use