Principaux documents
Z380334
Jars Nalgene® style 2117
capacity 1000 mL
Synonyme(s) :
laboratory jars, nalgene jars
1 PKG
88.80 CHF
4 PKG
355.00 CHF
88.80 CHF
Check Cart for Availability
Changer de vue
1 PKG
88.80 CHF
4 PKG
355.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
88.80 CHF
Check Cart for Availability
Matériaux
polymethylpentene bottle
polypropylene cap
polypropylene screw closure
round transparent bottle
Joint
Joints threaded neck (120)
Description
Autoclavable with closure completely unscrewed
Caractéristiques
with cap
Conditionnement
pkg of 4
Fabricant/nom de marque
Nalgene® 2117-1000
Diam. du flacon × H
112 mm × 151 mm
Capacité
1000 mL
Catégories apparentées
Description générale
Transparent, with good chemical resistance.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries