Principaux documents
Z362891
Sigma-Aldrich
Nalgene® benchtop β waste container
capacity 2,000 mL
Synonyme(s) :
lab waste receptical, liquid waste holder
1 EA
305.00 CHF
305.00 CHF
Date d'expédition estimée le28 mai 2025
Changer de vue
1 EA
305.00 CHF
About This Item
Code UNSPSC :
26142300
Nomenclature NACRES :
NB.35
305.00 CHF
Date d'expédition estimée le28 mai 2025
Matériaux
acrylic
Fabricant/nom de marque
Nalgene 6710-2000
Capacité
2,000 mL
Catégories apparentées
Description générale
3/8 in. thick, encases waste bottle without obstructing access. Shield cap fits securely with bottle closure in place. Provided with wide-mouth polyethylene bottle suitable for aqueous solution. Disposal of organic solvents may require a different inner bottle.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries