Principaux documents
Z362824
Nalgene® Polymethylpentene Erlenmeyer flask
capacity 1,000 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
2 EA
68.40 CHF
68.40 CHF
Check Cart for Availability
Changer de vue
2 EA
68.40 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
68.40 CHF
Check Cart for Availability
Matériaux
colorless polypropylene cap
flat bottom
polymethylpentene
wide-neck
Joint
Joints neck (53)
Stérilité
non-sterile
Caractéristiques
closure type Screw cap
with cap
Conditionnement
pack of 2
Fabricant/nom de marque
Nalgene 4109-1000
Capacité
1,000 mL
Catégories apparentées
Description générale
Excellent combination of autoclavability, chemical resistance, impact strength, and clarity. Transparent to microwave radiation. Polypropylene screw closure.
Autres remarques
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries