Principaux documents
Z325309
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 2 mL, two marks
12 EA
103.00 CHF
103.00 CHF
Check Cart for Availability
Changer de vue
12 EA
103.00 CHF
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
103.00 CHF
Check Cart for Availability
Matériaux
AR-Glas®
Description
partial delivery
Caractéristiques
class AS
Conditionnement
pack of 12 ea
Fabricant/nom de marque
BRAND 29723
Plage de mesure du volume
2 mL, accuracy: 0.010 mL
Capacité
2 mL
Catégories apparentées
Description générale
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Autres remarques
BR29723 (India)
Informations légales
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Protocoles
This page contains detailed information on working with and handling pipettes and pipetting aids.