Principaux documents
WHAWB120462
EasiCollect (50)
device for collection of buccal DNA samples, pkg of pack of 50 devices
Synonyme(s) :
Whatman FTA
50 EA
290.00 CHF
290.00 CHF
Check Cart for Availability
Changer de vue
50 EA
290.00 CHF
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.22
290.00 CHF
Check Cart for Availability
Caractéristiques
Included: A removable 2 × 2 in. (5 × 5 cm) indicating FTA card.
Fabricant/nom de marque
Qiagen WB120462
Conditionnement
pkg of pack of 50 devices
Description générale
EasiCollect device simplifies buccal cell collection and transportation. The design has been engineered for maximal cell collection and uniform sample application to FTA Cards to overcome individual differences. The pressure clips molded into the EasiCollect device ensure a uniform and constant pressure for reproducible transfer of cells to the FTA Card.
Caractéristiques et avantages
50 buccal sample collection devices for simplified sample uptake and transportation.
Autres remarques
Field of Use : For internal research use only
Protocoles
This document shows intructions for EasiCollect™ device from Cytiva.