Principaux documents
TMO3665-0025
Nalgene® economy graduated cylinder
PMP, volume 25 mL, case of 18 ea
Synonyme(s) :
PMP graduated cylinder, PMP graduated flask
18 EACH
180.00 CHF
180.00 CHF
Date d'expédition estimée le10 avril 2025
Changer de vue
18 EACH
180.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
180.00 CHF
Date d'expédition estimée le10 avril 2025
Matériaux
PMP
Conditionnement
case of 18 ea
Fabricant/nom de marque
Nalgene 3665-0025
Capacité
25 mL
Volume
25 mL
Description générale
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries