Yes, this product may be used to incubate slides at 65°C overnight in formamide. These Coplin Staining Jars are made from polypropylene which is very resistant to most chemicals, including dimethyl formamide. These jars may also be autoclaved at 121ºC. The manufacturer does not report a volume for this item, however the capacity is expected to be approximately 50 mL.
S5641
Scienceware® Coplin staining jar with screw cap
high-density polypropylene
Synonyme(s) :
coplin jar, microscope slide staining jar
146.00 CHF
About This Item
146.00 CHF
Produits recommandés
Matériaux
flat screw cap (53 mm in diameter)
high-density polypropylene
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art F44208-1000
Description générale
- Durable polypropylene construction
- Sealing ring makes jar watertight
- Stores up to 10 standard slides
- Autoclavable
Les clients ont également consulté
Articles
Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.
Protocoles
Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.
-
Can this product be used at 65°C overnight with slides incubated in a solution of formamide? And how much volume can this product hold?
1 answer-
Helpful?
-
-
Is it suitable for Xylene?
1 answer-
This product is made of polypropylene and is compatible with xylene at room temperature.
Helpful?
-
-
Can this product be used in microwave applications?
1 answer-
Yes, this product is suitable for microwave applications.