Principaux documents
HS120021
Totemax® Blood Collection Tray
blue
1 EACH
67.10 CHF
67.10 CHF
Check Cart for Availability
Changer de vue
1 EACH
67.10 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
67.10 CHF
Check Cart for Availability
Matériaux
blue
high-density polyethylene resin
Caractéristiques
Phlebotomy supplies and accessories
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Heathrow Scientific HS120021
L × l × H
432 mm × 279 mm × 279 mm
Description générale
The Totemax Blood Collection Tray′s unique phlebotomy tray design and patented hook feature allows the tray to be on or overmost bed rails, foot-boards and handrails outside a typical patient room. Keep the tray off the floor, work top or bed.
Caractéristiques et avantages
  • Made from food grade HDPE for a durable non-breakable design
  • Deep tray design helps reduce spills
  • Hooks lock in place when not in use
Informations légales
Totemax is a registered trademark of Heathrow Scientific LLC