Principaux documents
GE10600122
Amersham Hybond® P Western blotting membrane sandwich, PVDF
pore size 0.2 μm, sheet W × L 80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers, pkg of 10 ea
About This Item
Code UNSPSC :
41105339
Nomenclature NACRES :
NB.22
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Conditionnement
pkg of 10 ea
Fabricant/nom de marque
Cytiva 10600122
Feuille, l × L
80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers
Dimension de pores
0.2 μm pore size
Catégories apparentées
Description générale
Amersham Hybond P PVDF blotting membrane, with 0.2 µm pore size, are designed for use with chemiluminescent and fluorescent detection methods for proteins of > Mr 20 000.
Caractéristiques et avantages
  • Contains 10 Western blotting sandwiches, consisting of precut Amersham Hybond P 0.2 µm PVDF membranes preassembled with 2 × 3MM Chr Blotting papers.
Stockage et stabilité
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Informations légales
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Guoxia Zhu et al.
Neural plasticity, 2020, 8823785-8823785 (2020-10-22)
Sound conditioning (SC) is defined as "toughening" to lower levels of sound over time, which reduces a subsequent noise-induced threshold shift. Although the protective effect of SC in mammals is generally understood, the exact mechanisms involved have not yet been
Articles
This page describes troubleshooting strategies in the detection of GST-tagged proteins.
Protocoles
This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.