Principaux documents
GBL460003
Grace Bio-Labs press fit tubing connectors
pack of 20 ea
Synonyme(s) :
press fit fitting, tubing connectors
20 EA
39.40 CHF
39.40 CHF
Check Cart for Availability
Changer de vue
20 EA
39.40 CHF
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.51
39.40 CHF
Check Cart for Availability
Conditionnement
pack of 20 ea
Fabricant/nom de marque
Grace Bio-Labs 460003
Description générale
Press fit tubing connectors provide affordable and easy to use means to connect Grace′s chamber products to pumps and other devices for microfluidic applications.
Simply apply connectors by pressing the adhesive coated surface to the filling ports on the chamber surface and insert the appropriate length of tubing through the opening on the top. The simple hourglass design through the wall of the connector tightly secures the tubing to form a leak-proof seal in communication with the chamber. The open end of the tubing may be connected to a syringe or pump using a 25 gauge cannula or needle.