GBL204836
Grace Bio-Labs ProPlate® clips for microarray systems
stainless steel non-numbered, pack of 2 ea
Synonyme(s) :
microarray clips, stainless steel spring clips
23.60 CHF
About This Item
23.60 CHF
Produits recommandés
Caractéristiques
stainless steel non-numbered
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Grace Bio-Labs 204836
Catégories apparentées
Description générale
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
- Up to 64 leak-proof wells on any slide surface without the use of adhesive
- Microtiter plate well spacing facilitates use with multichannel pipettes
- 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes