Principaux documents
EP6346000021
Eppendorf® Mastercycler® Nexus Thermal Cyclers
SX1, AC/DC input 110 V, 50 - 60 Hz
About This Item
Code UNSPSC :
41102404
Code UNSPSC :
41102421
Nomenclature NACRES :
NC.21
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Caractéristiques
SX1
Fabricant/nom de marque
Eppendorf® 6346000021
Technique(s)
PCR: suitable
Catégories apparentées
Description générale
Flexlid Concept Due to the flexlid concept, the cycler can deal with all types of PCR consumables, from low volume all the way up to 0.5 mL PCR tubes. Of course, all common PCR plates fit – and if your throughput becomes even higher, you can just connect one or two Mastercycler nexus eco to your Mastercycler nexus
Product Information
With the versatile Mastercycler nexus you can choose the PCR consumables that best suit your assay. The PCR cycler nexus can accommodate 96-well PCR plates, 0.2 mL PCR tubes, 0.2 mL PCR tube strips and 0.5 mL PCR tubes. With a noise level of less than 40 dB[a], the nexus is very quiet and energy-efficient at the same time.
In the age of networking, the new Mastercycler nexus is your reliable companion when it comes to the daily routine of PCR. It is easy to use, does not need much space or energy and sends you an e-mail when it is done – what else do you need from a PCR cycler?
Certified Quality
All Eppendorf cyclers follow these quality guidelines:
  • Individual, documented quality control certificates
  • Calibration accordingly to national and international standards: NIST (USA), DKD/PTB (Germany), UKAS/NPL (UK)
  • UL/cUL listed
The Mastercycler nexus is a whisper quiet PCR machine. Low noise levels make working in your lab more pleasant and enjoyable, particularly when you are thinking about your next big breakthrough findings….
Application
Standard PCR, Fast PCR, PCR optimization
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG