EP5825719000
Adapter for Eppendorf® A-4-81 MTP/Flex
capacity, 1 x cell culture flask, pack of 2
223.00 CHF
About This Item
223.00 CHF
Produits recommandés
Caractéristiques
capacity 1 x cell culture flask
Holds 25 cm2 flasks
Conditionnement
pack of 2
Fabricant/nom de marque
Eppendorf® 5825719000 (Global)
Technique(s)
centrifugation: suitable
Compatibilité
for use with MTP/Flex bucket
Description générale
Caractéristiques et avantages
- Adapter to spin down cell cultures directly in a cell culture flask
- Suitable for 25 cm2 and selective 75 cm2 cell culture flasks
- Adapters can be used in Eppendorf plate rotor A-4-81-MTP/Flex