Principaux documents
EP3116000112
Accessories for Eppendorf® Pipettes & Repeaters
Pipette Holder 2, for one Eppendorf Research™, Eppendorf Research™ plus, Reference™, Reference™ 2 or Biomaster™
1 EA
38.40 CHF
38.40 CHF
Check Cart for Availability
Changer de vue
1 EA
38.40 CHF
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
38.40 CHF
Check Cart for Availability
Caractéristiques
for one Eppendorf Research, Eppendorf Research plus, Reference, Reference 2 or Biomaster
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Eppendorf® 3116000112 (Global)
Description générale
Accessories for Eppendorf pipettes and repeaters,
Eppendorf® provides holders suitable for all current manual and electronic liquid handling instruments as well as most predecessor manual pipettes. These holders allow you to have your pipette and Multipette at convenient locations, such as mounted to a wall, on a shelf above the bench, or inside a biological safety cabinet.
Caractéristiques et avantages
  • Compatible for one Eppendorf Research®, Eppendorf Research® plus, Eppendorf Reference®, Eppendorf Reference® 2 or Biomaster®
  • for Pipette Carousel 2 and Charger Carousel 2 or wall mounting, sticky tape is included for effortless installation
Informations légales
Biomaster is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.