Principaux documents
DWP96500Y4-EP
Eppendorf® Deepwell Plate 96/500μL
PCR Clean, volume 500 μL, yellow border with clear wells, pkg of 40 plates (5 bags x 8 plates)
About This Item
Code UNSPSC :
41122107
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Stérilité
non-sterile
Caractéristiques
PCR Clean
yellow border with clear wells
Conditionnement
pkg of 40 plates (5 bags x 8 plates)
Fabricant/nom de marque
Eppendorf® 951031828
Disponibilité
available only in North America
Technique(s)
PCR: suitable
Taille
96 well
Volume
500 μL
Puits
96
Description générale
Eppendorf® Deepwell Plates 96 and 384 are high-performance plates for all manual and automated applications – from sample storage at -86 ºC to DNA denaturation at 100 ºC. Manufactured from pure polypropylene (PP), these plates offer innovative features: More dependability. More efficiency. More stability. The plates are available with a white, yellow, green, or blue border. For easy identification of wells, a high-contrast alphanumeric label is laser-engraved in the border. This OptiTrack® matrix allows you to find your sample up to 30 % more quickly while reducing pipetting errors. With all of these sophisticated features, these plates set new standards in your laboratory.
Caractéristiques et avantages
  • OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • Minimal remaining volume and high uniformity from well to well
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Easy and reliable stacking
  • g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
  • High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
  • Autoclavable (121?°C, 20?min)
  • Available with barcode. Additional details available at: www.eppendorf.com/barcode
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG