CLS4519156
Corning® ProCulture® spinner flask accessories
side arm fitting, Dual with 1/4 inlets for 6 L spinner flask, Gas and Media Handling Fitting with Dual 1/4 in. Inlets for 6L Angled Sidearm Flasks
Synonyme(s) :
container accessory
609.00 CHF
Date d'expédition estimée le13 mai 2025
About This Item
609.00 CHF
Date d'expédition estimée le13 mai 2025
Fabricant/nom de marque
Corning 4519-156
Description générale
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.