Principaux documents
CLS4519150
Corning® ProCulture® spinner flask accessories
Media Handling Fitting with Dual 1/8 in. Inlets for 8L or 15L Vertical Sidearm Flasks
Synonyme(s) :
container accessory
1 EA
586.00 CHF
586.00 CHF
Check Cart for Availability
Changer de vue
1 EA
586.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
586.00 CHF
Check Cart for Availability
Fabricant/nom de marque
Corning 4519-150
Description générale
Side arm fittings used for a variety of functions in angled sidearm Spinner Flasks during cell culture. Sidearm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. Fittings comprise of a PET insert with a Viton O-ring and a polypropylene sealing cap & 316 stainless steel tubes.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Informations légales
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries