Principaux documents
CLS401025
Corning® LSE Power Adapter
AC/DC input 24 V
Synonyme(s) :
Corning® LSE Adapters
1 EA
56.90 CHF
56.90 CHF
Check Cart for Availability
Changer de vue
1 EA
56.90 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
SB.56
56.90 CHF
Check Cart for Availability
Fabricant/nom de marque
Corning 401025
Adéquation
suitable for (for use with Corning LSE compact centrifuge)
Description générale
These are optional adapters for Corning LSE compact centrifuge rotors: CLS480136, CLS480137, CLS480138; CLS480139
Informations légales
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.