Principaux documents
C079400
Sigma-Aldrich
Empty Synthesis Column
1 μmol (Colorless Body)
100 EA
215.00 CHF
215.00 CHF
Check Cart for Availability
Devis pour commande en gros
Changer de vue
100 EA
215.00 CHF
About This Item
Code UNSPSC :
12352200
Nomenclature NACRES :
NA.51
215.00 CHF
Check Cart for Availability
Devis pour commande en gros
Conditionnement
pkg of 1 × 100 ea
Compatibilité
configured for ABI
configured for PerkinElmer
Description générale
Empty Synthesis Columns are color coded and colorless empty column bodies with filters. C079300 empty synthesis column is a colorless body designed for ABI and Expedite Synthesizers.
Informations légales
ABI is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Expedite is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries