Principaux documents
BMSIPS2050PS1
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
flat blade, stainless steel
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.72
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Benchmark Scientific IPS2050-P-S1
Diam. × L
70 mm × 350 mm
Description générale
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
This one line, stainless steel propeller is useful for low viscosity, mid or high speed mixing.
Application
stirring, mixing