Principaux documents
BAF134810075
Bel-Art® Write-on Label Tape
roll size 15 yard, yellow, width 3/4 in., pk of 4
Synonyme(s) :
labelling tape, moisture resistant label tape, solvent resistant label tape
4 EA
37.90 CHF
37.90 CHF
Check Cart for Availability
Changer de vue
4 EA
37.90 CHF
About This Item
Code UNSPSC :
55121611
Nomenclature NACRES :
NB.35
37.90 CHF
Check Cart for Availability
Matériaux
yellow
Conditionnement
pk of 4
Fabricant/nom de marque
Bel-Art F13481-0075
Taille du rouleau
15 yard
Largeur
19.05 mm
3/4 in.
Description générale
Select from six tape colors that correspond with familiar hazard color codes to highlight safety information written on the labels. Use yellow for chemical reactivity, green for safety labels, blue indicates health hazards, red alerts to flammability; orange indicates a biohazard and white can describe a specific hazard. The tape is custom manufactured from proprietary materials that resist moisture and most solvents. Will adhere to PTFE fluoropolymer resin covered materials and any clean surface. A maximum temperature of 247 degrees C for 30 minutes can be endured.
Caractéristiques et avantages
  • No sticky residue lingers when tape is stripped off
  • Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP