AXYFXF20RS
Pipette filter tips for Beckman® FX Robotics System
racked, capacity 20 μL, pkg of 5x960tips/cs, sterile
Synonyme(s) :
pipet tips
About This Item
Produits recommandés
Stérilité
sterile
Fabricant/nom de marque
Axygen Inc FXF-20-R-S
Corning
Conditionnement
pkg of 5x960tips/cs
Capacité
20 μL
Volume
20 μL
Description générale
- Axygen® 96-well tips are optimized for use with Beckman Coulter, Biomek FX, Biomek NX, Biomek 3000, and Multimek AP-96, SPAN-8, MP20, MP200, and AP-384 head.
- Clear polypropylene tips are free of RNase-/DNase-/human DNA
- Features an aerosol-resistant filter.
- Automation tips are manufactured to stringent specifications and assembled via automation to ensure consistent performance and quality.