Principaux documents
Z413852
Sigma-Aldrich
Aldrich® reducing adapter
female joint: ST/NS 14/20, male joint: ST/NS 24/40
Synonyme(s) :
glass adapter, glass adapters
1 EA
45.60 CHF
45.60 CHF
Check Cart for Availability
Changer de vue
1 EA
45.60 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
45.60 CHF
Check Cart for Availability
Joint
female joint: ST/NS14/20
male joint: ST/NS24/40
Catégories apparentées
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC