Principaux documents
Z759295
Sigma-Aldrich
Joint clip, standard taper
joint: 14, yellow polyethylene
Synonyme(s) :
NS joint clamps, NS joint clips, National Standard joint clamps, National standard joint clips
50 EA
211.00 CHF
211.00 CHF
Check Cart for Availability
Changer de vue
50 EA
211.00 CHF
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
211.00 CHF
Check Cart for Availability
Matériaux
yellow polyethylene
Joint
joint: 14
Conditionnement
pkg of 50 ea
Description générale
Color coded, rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode; withstands temperatures up to 120 °C.
Les clients ont également consulté
Slide 1 of 2
1 of 2