Principaux documents
Z743189
Julabo® bath covers
with built in heat exchanger to fit CORIO CD-BC4/BC6
1 EA
433.00 CHF
433.00 CHF
Check Cart for Availability
Changer de vue
1 EA
433.00 CHF
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
433.00 CHF
Check Cart for Availability
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 9970240
Catégories apparentées
Description générale
Bath cover with built-in heat exchanger. To use the heat exchanger, a hose with an inner diameter of 10/12 mm is required.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.