Principaux documents
Z742518
Block for digital dry bath and MyBlock
Holds 24 x 0.5 mL tubes
1 EA
165.00 CHF
165.00 CHF
Check Cart for Availability
Changer de vue
1 EA
165.00 CHF
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NB.77
165.00 CHF
Check Cart for Availability
Caractéristiques
Holds 24 x 0.5 mL tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Benchmark Scientific BSW05
Description générale
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Informations légales
MyBlock is a trademark of Benchmark Scientific, Inc.